Imprimer
Masterflex® X-Rayable and Metal Detectable Sanitary Gaskets, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
30566-37_U 30566-35 30566-39
MFLA30566-37_UEA 0 EUR
MFLA30566-37_U MFLX30566-35 MFLX30566-39 MFLX30566-37 MFLX61010-25 MFLX61010-26 MFLX61010-27 MFLX61010-28 MFLX61010-29 MFLX61010-30
Masterflex® X-Rayable and Metal Detectable Sanitary Gaskets, Avantor®
Scellés Joints
Easily detect lost elastomeric fragments to prevent contamination and product recalls.
  • Designed for food processing, microbial, high-temperature, and mechanical applications
  • Available in five materials
These metal-detectable gaskets provide an easy and cost effective solution with metal detectable elastomers. By manufacturing a standard elastomer with a metal impregnated compound, displaced rubber material can now be located by an in-line metal detector.
As a result of excessive use, shearing and damage can occur to gaskets causing undetectableEn savoir plus