Imprimer
Flowline Switch-Tek guard RF capacitance level switch
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
43299-10
MFLX43299-10EA 471 EUR
MFLX43299-10
Flowline Switch-Tek guard RF capacitance level switch
Capteurs de niveau
Selectable normally open or normally closed relays increase system flexibility.
  • Connect up to three switches to one controller for maximum protection
SuperGuard(TM) RF capacitance level switch is the perfect switch for conductive, light-to-medium coating, or scaling liquids like soap solutions, printing ink, and wastewater. Polypropylene construction is ideal for corrosive environments. Includes 8-ft cable.