Imprimer
Masterflex® 3/4" Sanitary Clamp Medium Flow Female Quick-Disconnect Fittings, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX40257-00EA 159 EUR
MFLX40257-00 MFLX40257-01 MFLX40257-05 MFLX40257-06 MFLX40257-10 MFLX40257-11 MFLX40257-15 MFLX40257-16 MFLX40257-20 MFLX40257-21 MFLX40257-25 MFLX40257-26 MFLX40257-30 MFLX40257-31 MFLX40257-35 MFLX40257-36
Masterflex® 3/4" Sanitary Clamp Medium Flow Female Quick-Disconnect Fittings, Avantor®
Raccords et connecteurs
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Medium Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,En savoir plus