Imprimer
Masterflex® Single-Use Sanitary Gaskets, TPE, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX31201-64EA 51.7 EUR
MFLX31201-64 MFLX31201-65 MFLX31201-66
Masterflex® Single-Use Sanitary Gaskets, TPE, Avantor®
Scellés Joints
Ensure a tight seal in your sanitary fluid line.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Always in stock—most materials and configurations available to ship same day
  • Suitable for the most demanding biopharma fluid transfer applications
  • Reliable for sensitive fluid applications with low platelet adhesion and protein binding
Sanitary Tri-Clamp® gaskets secure the connection and prevent leakage from joining two sanitary fittings. TheseEn savoir plus