Imprimer
Plastomatic Sanitary Plastic Ball Valves
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
30532-01 30532-03 30532-05 30532-07 30532-17
MFLX30532-01EA 534 EUR
MFLX30532-01 MFLX30532-03 MFLX30532-05 MFLX30532-07 MFLX30532-17
Plastomatic Sanitary Plastic Ball Valves
Vannes Ball Valves
Precision machined and tested to ensure leak-free connections.
  • Standard Tri-Clamp connections are easy to plumb into sanitary fluid paths
  • Smooth flow path minimizes turbulence and pressure loss
  • Valve design permits full flow and pressure in either direction
These industry standard valves feature PTFE bearings and seats to virtually eliminate friction and minimize wear. Fully concentric polished ball further ensures leak-free performance. Available in polyproplyene (PP) or PVDF body material with EPDM seals.