Imprimer
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ 2001, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX06475-15EA 300 EUR
MFLX06475-15 MFLX06475-24
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ 2001, Avantor®
Tuyaux Tubes pour pompe péristaltique
Compatible with aggressive chemicals.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique
En savoir plus