Imprimer
Masterflex® Metal Metering Valves, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
03214-81 03214-82 03214-83 03214-85 03214-86 03214-91 03214-92 03214-93 03214-94 03214-95 03214-96 03218-76 03218-77 03218-84 03218-85 03214-84 03218-78 03218-79 03218-81 03218-83 03218-87
MFLX03214-81EA 115 EUR
MFLX03214-81 MFLX03214-82 MFLX03214-83 MFLX03214-85 MFLX03214-86 MFLX03214-91 MFLX03214-92 MFLX03214-93 MFLX03214-94 MFLX03214-95 MFLX03214-96 MFLX03218-76 MFLX03218-77 MFLX03218-84 MFLX03218-85 MFLX03214-84 MFLX03218-78 MFLX03218-79 MFLX03218-81 MFLX03218-83 MFLX03218-87
Masterflex® Metal Metering Valves, Avantor®
Vannes Metering Valves
Choose from different metal materials based on your application.
  • Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.