Imprimer
Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
03217-58 03217-63 03217-68 03217-74 03217-76 03217-79 03227-04 03227-08 03227-12 03227-14 03227-20 03227-24 03227-84 03229-05 03229-09 03229-13 03229-25 03295-04 03295-06 03295-08 03295-12 03295-14 03295-20 03295-22 03295-24 03295-26 03295-28
MFLX03217-53EA 478 EUR
MFLX03217-53 MFLX03217-57 MFLX03217-62 MFLX03217-67 MFLX03217-72 MFLX03217-73 MFLX03227-06 MFLX03227-10 MFLX03227-16 MFLX03227-18 MFLX03227-22 MFLX03227-26 MFLX03227-28 MFLX03227-30 MFLX03229-07 MFLX03229-11 MFLX03229-15 MFLX03229-17 MFLX03229-19 MFLX03229-21 MFLX03229-23 MFLX03229-27 MFLX03229-29 MFLX03229-31 MFLX03295-10 MFLX03295-16 MFLX03295-18 MFLX03217-58 MFLX03217-63 MFLX03217-68 MFLX03217-74 MFLX03217-76 MFLX03217-79 MFLX03227-04 MFLX03227-08 MFLX03227-12 MFLX03227-14 MFLX03227-20 MFLX03227-24 MFLX03227-84 MFLX03229-05 MFLX03229-09 MFLX03229-13 MFLX03229-25 MFLX03295-04 MFLX03295-06 MFLX03295-08 MFLX03295-12 MFLX03295-14 MFLX03295-20 MFLX03295-22 MFLX03295-24 MFLX03295-26 MFLX03295-28
Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Débitmètres
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,En savoir plus