Imprimer
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
TWS3060SU304S TWS3096SU304S TWS3660SU304S TWM3048SU304S TWM3096SU304S TWS3030SU304S TWS3036SU304S TWM3072SU304S TWM2436SU304S TWM2448SU304S TWM2460SU304S TWS2436SU304S TWS2448SU304S TWS2460SU304S TWM3030SU304S TWM3036SU304S
METOTWS3060SU304SEA 0 EUR
METOTWS3060SU304S METOTWS3096SU304S METOTWS3660SU304S METOTWM3048SU304S METOTWM3096SU304S METOTWS3030SU304S METOTWS3036SU304S METOTWM3072SU304S METOTWM2436SU304S METOTWM2448SU304S METOTWM2460SU304S METOTWS2436SU304S METOTWS2448SU304S METOTWS2460SU304S METOTWM3030SU304S METOTWM3036SU304S
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Mobilier Tables
TableWorx is a truly modular platform of performance work tables designed to allow you to customise your perfect solution.
All stainless series TableWorx performance work tables feature a 16-gauge, Type 304 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90° turned down edges on all four sides. All sizes are available with an optional 51 mm stainless steel backsplash, which is formed into the work surface.
All stainless series TableWorx performance work tables feature type 304 stainless steel legs and leg mounts. A type 304En savoir plus