Imprimer
Divider and tote boxes for shelving systems
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
DL93080NAT DS93080NAT MTB93030W MTB93060W DL93030NAT DS93030NAT MTB93080W
METODL93080NATEA 14.3 EUR
METODL93080NAT METODS93080NAT METOMTB93030W METOMTB93060W METODL93030NAT METODS93030NAT METOMTB93080W
Divider and tote boxes for shelving systems
Plateaux/Récipients/Pots Plateaux à tiroirs
The tote box provide safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers.
PP base resin.
  • Ideal for storage and can be transported
  • Available in various colour options
  • Injection molded for longer life and more uniform wall thickness
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable