Imprimer
Sampling swabs, NRSII™ Transwab®
Fournisseur: MEDICAL WIRE EQUIPMENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MW776
MEDWMW776EA 1390 EUR
MEDWMW776
Sampling swabs, NRSII™ Transwab®
Écouvillons Écouvillons d'échantillonnage
Preimmersed swabs for use with a self standing tube and a high visibility blue cap and swab, which is easy to find and retrive if accidentally dropped.
  • Blue cap and swab which can be easily retrived if dropped
  • Reduced packaging and storage space, ready to use
  • Ambient storage and transport
  • Precise fill for accurate assessment of contamination levels
  • Neutralisers to allow for the accurate recovery of residual bacteria on food contact surfaces
Specially recommended for sampling previously cleaned surfaces.
Certifications: ISO 18593 and other nationalEn savoir plus