Imprimer
Blades for cutters, STYROPOR, No. 79, 179 and 17940
Fournisseur: MARTOR ARGENTAX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
179.60 17940.60 79.60
MARO179.60EA 28.4 EUR
MARO179.60 MARO17940.60 233-0212
Blades for cutters, STYROPOR, No. 79, 179 and 17940
Rasoirs et lames
Non corrosive blade for mounting in safety knives and cutters.
  • Two-sided ground-edge
  • Double bevel edge-ground
Mountable in:
SECUNORM PROFI40
SECUNORM 380
SECUNORM 590
ARGENTAX TAP-O-MATIC