Imprimer
Safety knife, SECUMAX 145
Fournisseur: MARTOR ARGENTAX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
145001.12
MARO145001.12EA 52.8 EUR
MARO145001.12
Safety knife, SECUMAX 145
Couteaux
The SECUMAX 145 is a compact safety knife for routine cutting tasks. The two cutting edges are safely concealed to protect fingers.
  • Blade made of glass fibre reinforced plastic
  • Two in one blade head for cutting and piercing
The disposable cutting tool easily copes with single-layer cardboard, film and packing tape, for example.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable