Imprimer
Screw Caps with Dropping Pipette for Thread Bottles
Fournisseur: MARIENFELD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5459903
MARI5459903EA 141 EUR
MARI5459903
Screw Caps with Dropping Pipette for Thread Bottles
Capuchons Bouchons pour flacons
Screw caps with dropping pipette.
  • Made of white polypropylene
  • With white rubber teat
  • With dropping pipette with bent, spherical tip made of clear glass
Screw caps GL 18 for thread bottles.
Cdt: Minimum order quantity: 100 pieces each type.