Imprimer
Lames d'adhésion, HistoBond®+
Fournisseur: MARIENFELD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0810401 0810411 0810421 0810431 0810441 0810451
MARI0810401EA 49.2 EUR
MARI0810401 MARI0810411 MARI0810421 MARI0810431 MARI0810441 MARI0810451
Lames d'adhésion, HistoBond®+
Lames Lames adhésives
These adhesion slides are composed of silane treated surfaces and 90° ground edges.
  • Coloured marking areas on one end
  • Specially used for applications of colour coding
  • Improved identification of specimens
Certifications: IVD directive 98/79/EC