Imprimer
Colorimetric rapid tests, nickel, VISOCOLOR® ECO
Marques : MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
931040 931240
MANA931040EA 139 EUR
MANA931040 MANA931240
Colorimetric rapid tests, nickel, VISOCOLOR® ECO
Kits de tests environnementaux
In ammonia solution, nickel ions react with diacetyldioxime after oxidation with bromine to form a reddish-brown dye.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
Nickel can be present in industrial waste water. It occurs as the divalent ion or as a nickel complex.
Insoluble nickel compounds (e. g. nickel cyanide, nickel carbonate) and the nickel cyano complexes are not determined.
Suitable for sea water testing after dilution.