Imprimer
pH indicator papers, Duotest and Tritest
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
90301 90302 90303 90304 90311 90313 90501 90502 90510 90511 90305 90306 90312 90314 90315 90316
662-0235EA 56.5 EUR
662-0235 MANA90302 516-6011 MANA90304 MANA90311 MANA90313 662-0195 662-0203 662-0115 662-0116 MANA90305 MANA90306 MANA90312 MANA90314 MANA90315 MANA90316
pH indicator papers, Duotest and Tritest
Papiers test Bandelettes indicatrices de pH
By the combination of several indicators on one test paper, a better recognition between different pH values is achieved. This makes the correct pH reading easier.
  • Long term stable and protected against external influences due to plastic reel
  • No accessories needed, always ready to use
  • Reliable results due to easy and accurate read-off from colour scale
  • Several indicators for more precise readings
Duotest indicator papers combine two different indicator zones on a single strip. The zones are separated by a hydrophobic barrier, which effectivelyEn savoir plus