Imprimer
Lovibond® SD40 Multi Meter, Handheld
Fournisseur: Lovibond
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
720040
LOVI720040EA 254 EUR
LOVI720040
Lovibond® SD40 Multi Meter, Handheld
Compteurs électrochimiques pH-mètres et multi-paramètres
The SD40 Multi meter is a compact, easy to use handheld meter for the measurement of pH/Con/TDS/Salt/T.
  • Transparent lid for easy maintenance and storage
  • Ideal lid design for a stable stand
  • Display colours and symbols for easy use
  • New multi-parameter models for individual measurement requirements
  • Robust plastic case with hanger
With its waterproof housing, the Pocket Tester is ideal for regular checks in industrial plants or swimming pools.
Plus, the transparent lid not only provides a secure stand for the Pocket Tester in daily use, but also makesEn savoir plus