Imprimer
Latex tests
Marques : LIOFILCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
96147
LIOF96147EA 217 EUR
LIOF96147
Latex tests
Tests microbiologiques Tests sur les anticorps Ag-AK, Latex
Identification of the main pathogenic microorganisms directly from clinical specimens.
  • Save time and labor with direct testing
  • Results in 18 to 24 hours
  • Confirmation with latex agglutination tests
  • Clear and well defined colour changes
  • Easy interpretation