Imprimer
Flacons, fond plat, col étroit, avec rebord renforcé
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
07045088 07045028 07045037 07045049 07045058 07045070 07045073 07045079 07045082 07045085
LENZ07045088EA 525 EUR
LENZ07045088 LENZ07045028 LENZ07045037 LENZ07045049 LENZ07045058 LENZ07045070 LENZ07045073 LENZ07045079 LENZ07045082 LENZ07045085
Flacons, fond plat, col étroit, avec rebord renforcé
Ballons Ballons à fond plat
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck diameter of 65 mm have a reinforced rims.
Certifications: Acc. to DIN ISO 1773 and in addition to standard.