Imprimer
Freeze dryer, console, FreeZone®
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
700612030 700612130 700612230 700612330 700621030 700611030 700611130 700611230 700611330 700621130 700622030 700622130 701211030 701211130 701211230 701212030 701212130 701212230 701221030 701221130 701222030 701222130 701811030 701811130 701811230 701812030 701812130 701812230 701821030 701821130 701822030 701822130 710611030 710611130 710611230 710612030 710612130 710612230 710621030 710621130 710622030 710622130 711211030 711211130 711211230 711212030 711212130 711212230 711221030 711222130 711221130 711222030
LBCP700612030EA 19670 EUR
LBCP700612030 LBCP700612130 LBCP700612230 LBCP700612330 LBCP700621030 LBCP700611030 LBCP700611130 LBCP700611230 LBCP700611330 LBCP700621130 LBCP700622030 LBCP700622130 LBCP701211030 LBCP701211130 LBCP701211230 LBCP701212030 LBCP701212130 LBCP701212230 LBCP701221030 LBCP701221130 LBCP701222030 LBCP701222130 LBCP701811030 LBCP701811130 LBCP701811230 LBCP701812030 LBCP701812130 LBCP701812230 LBCP701821030 LBCP701821130 LBCP701822030 LBCP701822130 LBCP710611030 LBCP710611130 LBCP710611230 LBCP710612030 LBCP710612130 LBCP710612230 LBCP710621030 LBCP710621130 LBCP710622030 LBCP710622130 LBCP711211030 LBCP711211130 LBCP711211230 LBCP711212030 LBCP711212130 LBCP711212230 LBCP711221030 LBCP711222130 LBCP711221130 LBCP711222030
Freeze dryer, console, FreeZone®
Lyophilisateurs
FreeZone® console freeze dryers with or without stoppering tray dryers are designed for lyophilising moderate to large loads and stoppering vials or serum bottles under vacuum. The console features a front viewing window and interior space for a vacuum and accommodation for a full size tray dryer such as the FreeZone® stoppering tray dryer pump (sold separately).
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ end
En savoir plus