Imprimer
Glove boxes, stainless steel combination, Protector®
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5080172 5080170 5080165 5080167
LBCP5080172EA 109750 EUR
LBCP5080172 LBCP5080170 LBCP5080165 LBCP5080167
Glove boxes, stainless steel combination, Protector®
Boîtes à gants et sacs à gants
These glove boxes with stainless steel liner provide inlet and outlet ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials and perform work with contamination-sensitive materials under vacuum or in an inert environment. Available with two or four glove ports (double version).
  • LCD display, large viewing area (double glove boxes have two viewing areas), space-saving inner and outer transfer chamber doors that pivot upward
  • Two internal valves to convert the glove box from a flow-through
En savoir plus