Imprimer
Fume absorbers
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3955221 3955421 3955420
LBCP3955221EA 0 EUR
LBCP3955221 LBCP3955421 LBCP3955420
Fume absorbers
Hottes et coffrets Hottes filtrantes
These fume adsorbers are for light duty histology cover slipping and staining areas. Activated carbon media adsorb organic odours and noxious vapors released by solvents like xylene and toluene while clean air is recirculated into the laboratory. These enclosures are low profile, completely portable and require no ductwork. They easily accommodate shallow countertops or those with overhanging cabinets.
  • Built-in, motorized impeller to provide face velocity up to 35 fpm
  • Glacier white, dry powder, epoxy-coated steel housing with perforated baffle for uniform
En savoir plus