Imprimer
Polyethylene Heavy-Duty Graduated Buckets
Fournisseur: LAMAPLAST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
161 162 163 165 164
LAMA161EA 13.1 EUR
LAMA161 LAMA162 LAMA163 LAMA165 LAMA164
Polyethylene Heavy-Duty Graduated Buckets
Godets
Made with low density polyethylene (LDPE), in white or black colour options.
  • Conical shape, litre graduations
  • With ergonomic handle
  • Food contact material approved
Buckets feature a flat bottom design to ensure total emptying and allow for easy cleaning.
Certifications: EU 10/2011.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable