Imprimer
Trypsin
Marques : Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
L0910-100 L0930-100 L0930-500 L0940-100 L0940-500 P0940-1GR P0940-100GR X0915-100 X0930-100 P5957-100GR L0909-100 L0931-100 L0931-500 L0932-100 X0920-100 L0941-100
L0910-100EA 12.2 EUR
L0910-100 L0930-100 L0930-500 L0940-100 L0940-500 P0940-1GR P0940-100GR X0915-100 X0930-100 P5957-100GR L0909-100 L0931-100 L0931-500 L0932-100 X0920-100 L0941-100
Trypsin
Suppléments et additifs pour culture cellulaire
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.