Imprimer

Penicillin/Streptomycin/Glutamine 100X

Marques : Biowest
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Penicillin/Streptomycin/L-Glutamine

L0014-100
L0014-100EA 25.5 EUR
L0014-100
Penicillin/Streptomycin/Glutamine 100X
Penicillin/Streptomycin/Glutamine

Glutamine penicillin streptamycin acts against gram-negative and gram-positive bacteria.


  • Storage temperature of –20 °C
  • Shelf life of 24 months
  • Endotoxin: <10 EU/ml


Used in cell culture applications at 10 ml/l. This concentration is for tissue culture media containing serum; serum-free media generally require lower concentration.


Attention: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR