Imprimer

Amphotericine B

Marques : Biowest
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

L0009-020 L0009-050 L0009-100
L0009-020EA 15.3 EUR
L0009-020 L0009-050 L0009-100
Amphotericine B
Amphotericine B

Amphotericin B used in tissue culture to prevent growth of yeasts and fungi. It changes the function and integrity of eucaryotic cell membranes by forming complexes with sterols (cholesterol) hence having no effect on bacteria. Cause leakage of glucose.


  • Yellow coloured, with a pH of 10 – 11,5
  • Shelf life of 24 month


Interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out.


Attention: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Formule: C₄₇H₇₃NO₁₇
Poids moléculaire: 924.08 g/mol
Numéro MDL: MFCD00877763
Numéro CAS: 1397-89-3

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR