Imprimer
Induction cooking plate
Fournisseur: KOEHLER TECHNISCHE PRODUKTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2-3874
KOEH2-3874EA 646 EUR
KOEH2-3874
Induction cooking plate
Appareils chauffants Plaques chauffantes
This induction cooking plate allows heating and cooking in a fast and effective way.
  • No heating time
  • Immediate power transmission
  • Easy to clean
  • Automatic shutdown
  • Control through touch control sensors
The plate has a Schott Ceran® cooking surface with a cooking zone of 100 – 200 nm and a heating zone of 80 to 200 mm diameter.