Imprimer
Sample bags, Whirl-Pak®
Fournisseur: KOEHLER TECHNISCHE PRODUKTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1-7136
KOEH1-7136EA 164 EUR
KOEH1-7136
Sample bags, Whirl-Pak®
Sachets Sachets d'échantillonnage
Whirl-Pak plastic bags are made up of low density polyethylene (LDPE) which is suitable for liquid samples for food, water and waste water testing and also for biological or chemical samples.
  • Strong welding seam
  • Lodged with a metal strap for opening and closing of the bag
  • White inscription pad
  • Sterile bags with closure
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904