Imprimer
Laboratory reactor systems, Reactron®
Fournisseur: KINEMATICA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
14090010 14090011 14090010-UK 14090020 14090020-UK 14090021 14090030 14090030-UK 14090031
KINE14090010EA 0 EUR
KINE14090010 KINE14090011 KINE14090010-UK KINE14090020 KINE14090020-UK KINE14090021 KINE14090030 KINE14090030-UK KINE14090031
Laboratory reactor systems, Reactron®
Systèmes de réacteurs de laboratoire
REACTRON® laboratory systems consist of process vessels (single and multiple wall, standard and customer-specific designs), stirring systems for macro-mixing with anchor stirrer), POLYTRON®/MEGATRON® homogenisers and mixing systems for crushing and micro-mixing, pipework, process controls and accessories, in alignment with the customer, product and process.
  • Single wall or double wall for the optimum application temperature
  • Borosilicate, glass or steel
  • Range of dispersers drives and generators
  • Simple change of aggregate
RT 1 standard systems: JacketedEn savoir plus