Imprimer
Cold trap, type CP
Fournisseur: KGW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1707
KGWA1707EA 9110 EUR
KGWA1707
Cold trap, type CP
Pièges à vide
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents and these cold traps can be used for many different tasks. The most common application is gas cleaning, drying and condensation process. Model CP 1 has got an additional vacuum flange NW 16 with a vacuum gauge.
  • Compact design
  • Easy handling
  • Frame work on wheels
  • Suitable for aggressive mediums
  • Table board made of stainless steel
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.