Imprimer
Dishes, crystallising
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
632411625115 632411625190 632411625230 632411624140 N632412635150 632411625095 632411625080 N632412624160 632411624080_U 632411625140 N632412624140 N632412624150 N632412624170 N632412635140 632411624115 632411624190 632411624230 632411624080 632411624095
KAVA632411625115EA 336 EUR
KAVA632411625115 KAVA632411625190 KAVA632411625230 KAVA632411624140 KAVAN632412635150 KAVA632411625095 KAVA632411625080 KAVAN632412624160 KAVA632411624080_U KAVA632411625140 KAVAN632412624140 KAVAN632412624150 KAVAN632412624170 KAVAN632412635140 KAVA632411624115 KAVA632411624190 KAVA632411624230 KAVA632411624080 KAVA632411624095
Dishes, crystallising
Plateaux/Récipients/Pots Cristallisoirs
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications.