Imprimer
Dishes, evaporating, flat or round Bottom, with spout
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
632411629060 632411604050 632411604100 632411604130 632411604160 632411604200 632411604230 632411629115 632411629095 632411629140 632411604060 632411629070
216-1192EA 12.1 EUR
216-1192 KAVA632411604050 KAVA632411604100 KAVA632411604130 KAVA632411604160 KAVA632411604200 KAVA632411604230 KAVA632411629115 KAVA632411629095 KAVA632411629140 KAVA632411604060 KAVA632411629070
Dishes, evaporating, flat or round Bottom, with spout
Plateaux/Récipients/Pots Capsules d'évaporation
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications. With flat bottom or rund bottom.