Imprimer
Dishes, crystallising
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
639 630 634
KART639EA 77.9 EUR
KART639 KART630 KART634
Dishes, crystallising
Plateaux/Récipients/Pots Cristallisoirs
Crystallising dishes, made of PTFE.
  • Chemical, mechanical and heat resistance
  • Used safely for continuous service from −270 up to +250 °C
Suitable for dissolutions, digestions with acids and corrosive agents and suitable for food stuffs.