Imprimer
Microtubes à centrifuger
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
279
KART279EA 37.1 EUR
KART279
Microtubes à centrifuger
Tubes Microtubes à centrifuger
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Flat cap with writing area
Ideal for samples analysis. Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to 11000×g.