Imprimer
Urine Bottles, Square
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0047900 0048000
KART0047900EA 259 EUR
KART0047900 KART0048000
Urine Bottles, Square
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons d'urine
Bottle for clearance testing on urine collected during the whole day long.
  • Rectangular base for easy storage and enhanced stability in use
  • Ergonomic handgrip allows easy pouring
Adapted to contain a human biological sample (urine) in order to perform diagnostic laboratory analysis.
Certifications: CE marked.