Imprimer
Hydraulic laboratory jacks
Fournisseur: JUCHHEIM Laborgerate GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
VL02295 VL02294 VL02296 VL02297 VL02297/A VL02297/AELOX. VL02297/A-K VL02297/E VL02297/G VL02297/GA VL02297/S
JUCHVL02295EA 6150 EUR
JUCHVL02295 JUCHVL02294 JUCHVL02296 JUCHVL02297 JUCHVL02297/A JUCHVL02297/AELOX. JUCHVL02297/A-K JUCHVL02297/E JUCHVL02297/G JUCHVL02297/GA JUCHVL02297/S
Hydraulic laboratory jacks
Crics de laboratoire
These hydraulic lifting stages feature a robust construction in light alloy or stainless steel.
  • Aluminium alloy or stainless steel
  • Available with hand or pedal operation