Imprimer
Positive pressure respirators, PowerCap® Active™
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CAE602-941-100 CAU601-001-100 CAU610-000-010 CAU340-001-100 CAU330-001-100 CAU080-000-000 CAU350-000-000 CAU071-001-100 CAU150-000-000 CAU030-000-000 CAE601-941-100
JSPACAE602-941-100EA 1510 EUR
JSPACAE602-941-100 JSPACAU601-001-100 JSPACAU610-000-010 JSPACAU340-001-100 JSPACAU330-001-100 JSPACAU080-000-000 JSPACAU350-000-000 JSPACAU071-001-100 JSPACAU150-000-000 JSPACAU030-000-000 JSPACAE601-941-100
Positive pressure respirators, PowerCap® Active™
Protection respiratoire Systèmes de respiration
The PowerCap® Active™ is a lightweight positive pressure respirator with eye and face protection, as well as EN812 bumpcap protection. Designed to protect the user from dust and most airborne particulate pollutants to EN12941 TH1P.
  • Lightweight, firm fitting, ergonomically designed positive pressure respirator
  • No face fit required
  • Cap uses JSP Hardcap A1+™ bump cap technology
  • Each battery lasts 8 hours
  • Polycarbonate face shield
  • Foam face seal
PowerCap® Active™ is designed to protect the user from dust and most airborne particulate pollutants to EN 12941En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable