Imprimer
Frittés encastrés PEEK
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
JR-1104-2P-5 JR-1104-05P-5 JR-1103-2P-5 JR-1103-05P-5 JR-1102-2P-5 JR-1102-05P-5 JR-1101-2P-5 JR-1101-05P-5 JR-1100-2P-5 JR-1100-05P-5 JR-1110-2P-5 JR-1110-05P-5 JR-1127-05P-5 JR-1125-2P-5 JR-1125-05P-5 JR-1111-5P-5 JR-1111-2P-5 JR-1111-05P-5 JR-1110-5P-5 JR-1125-5P-5 JR-1125-10P-5 JR-1128-5P-5
JOURJR-1104-2P-5EA 174 EUR
JOURJR-1104-2P-5 JOURJR-1104-05P-5 JOURJR-1103-2P-5 JOURJR-1103-05P-5 JOURJR-1102-2P-5 JOURJR-1102-05P-5 JOURJR-1101-2P-5 JOURJR-1101-05P-5 JOURJR-1100-2P-5 JOURJR-1100-05P-5 JOURJR-1110-2P-5 JOURJR-1110-05P-5 JOURJR-1127-05P-5 JOURJR-1125-2P-5 JOURJR-1125-05P-5 JOURJR-1111-5P-5 JOURJR-1111-2P-5 JOURJR-1111-05P-5 JOURJR-1110-5P-5 JOURJR-1125-5P-5 JOURJR-1125-10P-5 JOURJR-1128-5P-5
Frittés encastrés PEEK
Filtres Filtres de précolonnes
Made of stainless steel or titanium. Titanium frits are resistant to corrosion, biocompatible and recommended for protein analysis.
  • Intended for use with HPLC columns or in-line filters
  • Eliminate poorly swept volume and improves peak symmetry
The compressed PEEK ring forms a seal between the bottom of the fitting and the column end, preventing the mobile phase and the sample from entering the previously accessible poorly swept areas.