Imprimer

Ampicilline sel de sodium

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): D(-)-α-Aminobenzylpénicilline sel de Sodium

J63807.06 J63807.09
J63807.06EA 96.3 EUR
J63807.06 J63807.09
Ampicilline sel de sodium
Ampicilline sel de sodium

CAS No.: 69-52-3 Molecular Formula: C16H18N3NaO4S Formula Weight: 371.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 215° Solubility: Freely soluble in water. Sparingly soluble in acetone Beilstein Reference: 4119211 Merck Reference: 14,586 MDL No.: MFCD00064313

Formule: C₁₆H₁₈N₃NaO₄S
Poids moléculaire: 371.39 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00064313
Numéro CAS: 69-52-3
EINECS: 200-708-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR