Imprimer
Dry vacuum pumps, Dryfast®
Fournisseur: Welch by Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2019C-02
ILMV2019C-02EA 770 EUR
ILMV2019C-02
Dry vacuum pumps, Dryfast®
Pompe Pompes à vide à membrane
Pumps are constructed with PTFE heads, perfluoroelastomer valves and a PTFE coated molded diaphragm. Includes intake/exhaust hose barbs. DRYFAST® models include tunable vacuum control.
  • Chemical resistant
  • Oil free