Imprimer
Laboratory vacuum systems
Fournisseur: Welch by Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
115037
ILMV115037EA 4050 EUR
ILMV115037
Laboratory vacuum systems
Pompe Pompes à vide pour évaporateurs rotatifs
Compact, dry-running, chemical resistant pump systems with a range of vacuum control options that plays an ideal solution for many applications in chemical laboratories and research.
  • Both manual and automatic pump system with built-in solvent library
  • Close to 100% solvent recovery
  • Optimal stability with multi-lingual digital display
  • Maintenance-free drive system and proven long diaphragm life
  • Quick and easy maintenance for user-friendly operation
  • Inlet separator to protect pump from liquid and particle ingestion
  • Exhaust condenser for optimal solvent
En savoir plus