Imprimer
Magnetic stirrer, Topolino mobil
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3381300 3381302
IKAI3381300EA 379 EUR
IKAI3381300 IKAI3381302
Magnetic stirrer, Topolino mobil
Agitateurs Agitateurs magnétiques
Mini magnetic stirrer with durable brushless motor for mixing small quantities.
  • Continuously adjustable speed range
  • Powerful magnets for strong magnetic coupling
  • Uses standard replaceable AA rechargeable batteries or can be used with mains power
IP protection class according to DIN EN 60529: IP 20
Certifications: IP protection class according to DIN EN 60529: IP 20