Imprimer
Mechanical overhead stirrers, RKG-00-Bo, TURBOTRON®, IKA®
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
U098950 U084463 U087228
IKAAU098950EA 0 EUR
IKAAU098950 IKAAU084463 IKAAU087228
Mechanical overhead stirrers, RKG-00-Bo, TURBOTRON®, IKA®
Agitateurs Agitateurs à tige
Powerful, mechanically controlled agitators for intensive mixing with approval for use in Ex-Zone 1 (2G), temperature class T3 acc. to the ATEX 95 directive.
  • Suitable for open or pressureless vessels
  • Exchangeable stirring tools
Informations de commande: Supplied without cables.