Imprimer
Immersion Circulators, ICC 100 lite
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
20105647
IKAA20105647EA 1040 EUR
IKAA20105647
Immersion Circulators, ICC 100 lite
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation. ICC 100 lite is suitable for tempering liquids up to 100 °C and internal circulation.
  • USB and RS232 interface for monitoring, control and documentation
Increased safety: ICC 100 lite is approved for use with flammable liquids in accordance with DIN 12876, safety class III.
Convenient handling: The integrated stand enables easy cleaning and prevents important components fromEn savoir plus