Imprimer
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
11RIST2050
ICNA11RIST2050EA 99 EUR
ICNA11RIST2050
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Tampons Tampons d'électrophorèse Tampons d'hybridation
SSC buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates. SSC is used as a hybridisation buffer in immunoblotting and DNA microarray protocols.