Imprimer
DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
117022110
ICNA117022110EA 335 EUR
ICNA117022110
DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
MPure™ Forensic DNA Extraction Kit is used with the MPure-12™ aNAP system for the extraction and purification of genomic DNA from forensic samples.