Imprimer

Anti-IgG Rat Antibody

Fournisseur: MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

08641481
ICNA08641481EA 939 EUR
ICNA08641481
Anti-IgG Rat Antibody
Anticorps
The Immunoglobulin was isolated chromatographically from normal rat serum using DEAE and gel filtration techniques. The immunoglobulin underwent Protein G purification.

  • Can be reconstitute with 0,135 M NaCl.

It is useful for structural and biological studies, as a reference standard, or as starting material for immunogen preparation. It may be used as a reference antigen, standard, blocking agent, or coating protein in a variety of Immunoassays.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR