Imprimer

Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))

Fournisseur: MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

0855391
ICNA0855391EA 533 EUR
ICNA0855391
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Anticorps
Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to guinea pig IgG (whole molecule) and buffer salts. The total protein is measured using the Biuret procedure with bovine albumin as the standard. To ensure that the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with guinea pig IgG. The product is tested for purity and specificity at final concentration by immunoelectrophoresis. The antibody is predominantly goat IgG; no trace of albumin is detected.

Type: Secondary
Antigen: IgG
Clonality:
En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR