Imprimer
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0219989910 0219989925 0219989950 0219989980 0219989991
ICNA0219989910EA 77.2 EUR
ICNA0219989910 ICNA0219989925 ICNA0219989950 ICNA0219989980 ICNA0219989991
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Protéines et Peptides
AlbumiNZ™ Low Fatty Acid BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. This albumin has very low endotoxin and low free fatty acids, is tested for the absence of viruses according to 9CFR and has the highest possible biosafety level.
  • Off-white powder
  • Dissolves in ≤20 min (20% w/w in water)
AlbumiNZ™ Low Fatty Acid BSA supports growth of stem cells and increases protein production from hybridoma cells.